missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences WHEATON™ CompletePAK™ Clear Kits

Product Code. 17972591 Shop All Wheaton Science Products Products
Click to view available options
Volume (Metric):
2 mL
5 mL
10 mL
Closure Type:
OmniFlex 3G
OmniFlex 3G Igloo Lyo Stopper
OmniFlex 3G Serum Stopper
Ultra Pure
Ultra Pure Serum Stopper
Closure Color:
Blue
Red
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 17972591

Brand: DWK Life Sciences W991012CP

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

The WHEATON™ CompletePAK™ product line is a range of crimp top vials, rubber stoppers and seals that are sterilized ready-to-use and are off- the-shelf, and can be customized for any application reducing your supply chain due to our one source solution capability.

  • Sterilized, off-the-shelf, ready-to-use (RtU) products
  • Variety of Kitted and stand-alone sterile components – a solution for any application
  • Kitted solutions feature equal amounts of RtU components for reduced scrap rates
  • Complete manufacturing & certificate of analysis traceability
  • USP 788/EP 2.9.19 Particulate Matter in Injections, USP 85/EP 2.6.1, The Bacterial Endotoxins Test and USP 71/ EP 2.6.14 The Sterility Test, USP 660/EP 3.2.1 Glass Containers for Pharmaceutical Use, and USP 381/EP 3.2.9 Elastomeric Closures used in Parenteral Preparations
  • CompletePAK™ ModPAK Bulk configurations
  • Ready-to-Use, Just-in-Time, minimum 3 month shelf life from time of shipment
  • Container Closure Integrity (CCI) certification available upon request

Certifications

USP <788>, <85> and <71>; Container Closure Integrity (CCI) certified

TRUSTED_SUSTAINABILITY

Specifications

Closure Color Blue
Color Clear
Sterility Sterile
Volume (Metric) 10 mL
Includes Vial, Stopper, and Seal
No. per Case 104/Cs.
Product Line Wheaton CompletePAK
Closure Type Ultra Pure Serum Stopper
Certifications/Compliance USP 788, 85, and 71, Container Closure Integrity (CCI) certified
Closure Size 20 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.