missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences WHEATON™ CompletePAK™ Clear Kits
Shop All Wheaton Science Products Products
missing translation for 'orderingAttributeHoverText'
Volume (Metric):
2 mL
5 mL
10 mL
Closure Type:
OmniFlex 3G
OmniFlex 3G Igloo Lyo Stopper
OmniFlex 3G Serum Stopper
Ultra Pure
Ultra Pure Serum Stopper
Closure Color:
Blue
Red
missing translation for 'unitSize'
1 set
Description
- Sterilized, off-the-shelf, ready-to-use (RtU) products
- Variety of Kitted and stand-alone sterile components – a solution for any application
- Kitted solutions feature equal amounts of RtU components for reduced scrap rates
- Complete manufacturing & certificate of analysis traceability
- USP 788/EP 2.9.19 Particulate Matter in Injections, USP 85/EP 2.6.1, The Bacterial Endotoxins Test and USP 71/ EP 2.6.14 The Sterility Test, USP 660/EP 3.2.1 Glass Containers for Pharmaceutical Use, and USP 381/EP 3.2.9 Elastomeric Closures used in Parenteral Preparations
- CompletePAK™ ModPAK Bulk configurations
- Ready-to-Use, Just-in-Time, minimum 3 month shelf life from time of shipment
- Container Closure Integrity (CCI) certification available upon request
Certifications
USP <788>, <85> and <71>; Container Closure Integrity (CCI) certified
Spécification
Spécification
| Closure Color | Blue |
| Color | Clear |
| Sterility | Sterile |
| Volume (Metric) | 2 mL |
| Includes | Vial, Stopper, and Seal |
| No. per Case | 278/Cs. |
| Product Line | Wheaton CompletePAK |
| Closure Type | OmniFlex 3G |
| Certifications/Compliance | USP 788, 85, and 71, Container Closure Integrity (CCI) certified |
| Closure Size | 13 mm |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu