Learn More
DWK Life Sciences WHEATON™ CompletePAK™ Sterile Components Seals
The WHEATON™ CompletePAK™ product line is a range of crimp top vials, rubber stoppers and seals that are sterilized ready-to-use and are off- the-shelf, and can be customized for any application reducing your supply chain due to our one source solution capability.
Brand:  DWK Life Sciences W988009-S
 View more versions of this product
Product Code. 17932681
  • 790.00 € / Pack of 285
Explore more special offers
Description
Description
  • Sterile
  • Flip-off Seal
Certifications
USP <788>, <85> and <71>; Container Closure Integrity (CCI) certified
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Red
Sterile
Wheaton CompletePAK
USP 788, 85, and 71, Container Closure Integrity (CCI) certified
2 mL Serum Vial
Aluminum
285/Cs.
Sterile Flip Seal
13 mm