Learn More
DWK Life Sciences WHEATON™ CompletePAK™ Sterile Components Vials
The WHEATON™ CompletePAK™ product line is a range of crimp top vials, rubber stoppers and seals that are sterilized ready-to-use and are off- the-shelf, and can be customized for any application reducing your supply chain due to our one source solution capability.
992.00 € - 1385.00 €
Specifications
Sterility Sterile
Product Line Wheaton CompletePAK
View More Specs
Products 3
Product Code Brand Color Volume (Metric) Price Quantity & Availability  
Product Code Brand Color Volume (Metric) Price Quantity & Availability  
16297280
View Documents
DWK Life Sciences
W986000-S
Clear 2 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16207290
View Documents
DWK Life Sciences
W986001-S
Clear 5 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16217290
View Documents
DWK Life Sciences
W986002-S
Clear 10 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • Sterilized, off-the-shelf, ready-to-use (RtU) products
  • Variety of Kitted and stand-alone sterile components – a solution for any application
  • Complete manufacturing & certificate of analysis traceability
  • USP 788/EP 2.9.19 Particulate Matter in Injections, USP 85/EP 2.6.1, The Bacterial Endotoxins Test and USP 71/ EP 2.6.14 The Sterility Test, USP 660/EP 3.2.1 Glass Containers for Pharmaceutical Use, and USP 381/EP 3.2.9 Elastomeric Closures used in Parenteral Preparations
  • Ready-to-Use, Just-in-Time, minimum 3 month shelf life from time of shipment
  • Amber versions for light sensitive content packaging
Specifications
Specifications
Sterile
Wheaton CompletePAK