missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences™ Wheaton™ CryoELITE™ Vial rack

Benchmate rack for use with both freestanding and round bottom vials.

Brand:  DWK Life Sciences™ W985810

Product Code. 15336534

  • 231.00 € / Pack of 5
Estimated Shipment: 22-09-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Stackable for easy storage
  • Capacity of 50 cryogenic vials
  • Rack manufactured from polypropylene
  • Easily cleaned in an automatic washer or autoclavable at 121°C for 20 minutes
  • Non-skid feet offer stability for bench work
  • Each well has an inner diameter of 12.5mm with and overall rack footprint of 190 x 100 x 22mm
TRUSTED_SUSTAINABILITY
Specifications

Specifications

190 x 100 x 22 mm
50 Cryo Vials (12.5mm diameter)
Autoclavable (121°C for 20 min.)
Cryogenic Vial Rack
Freestanding and round bottom cryovials
5/cs.
Polypropylene
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DWK Life Sciences™ Wheaton™ CryoELITE™ Vial rack > CryoELITE Benchmate vial rack

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.