missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences™ Wheaton™ CryoFile™ and CryoFile XL™ Storage Boxes
For easy Identification and storage of cryogenic vials
Brand: DWK Life Sciences™ W651604
This item is not returnable.
View return policy
Description
- Accommodates 1.2 - 2mL vials and XL boxes store 5 mL Partitioned carton numbered from 1-81 keeps vials organized and easy to identify
- Numbering system printed on box lid and bottom of box provides easy indexing
- Water repellent allow for longer durability
- Withstands cryogenic temperatures to -196°C

Specifications
Blue | |
2 in. | |
5.1 in. | |
5.1 in. | |
For easy identification and storage of cryogenic vials | |
1 to 81 Vials | |
CryoFile™ Storage Box |
5.3 cm | |
13 cm | |
13 cm | |
5.1 x 5.1 x 2 in. (13 x 13 x 5.3 cm) | |
1.2 and 2mL vial | |
Corrugated Chipboard |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DWK Life Sciences™ Wheaton™ CryoFile™ and CryoFile XL™ Storage Boxes > For 1.2 and 2mL vials; Blue; 130L x 130W x 52mm H
Spot an opportunity for improvement?Share a Content Correction