Learn More
DWK Life Sciences Wheaton™ Deck Kit for Compact Roller System for Mini Bottles
Mini Bottle Deck Kit can be added to Mini Bottle Roller Culture Apparatus to accommodate an additional four bottles per deck.
Brand:  DWK Life Sciences 348920
 View more versions of this product
Product Code. 12458952
  • 1055.00 € / Each
Estimated Shipment: 10-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Can add up to a maximum of 2 kits per unit.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Deck Kit
4 kg
Mini Bottle Roller Culture Apparatus
9 lb.
Black
Assembled roller deck, 4 support posts, drive belt, necessary hardware and assembly instructions