missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Dounce Tissue Grinders
Ideal for enzyme studies. Wheaton™ Dounce Tissue Grinders are designed to retain a high percentage of cell nuclei, mitochondria, and proteins in soft tissues or cell suspensions.
Brand: DWK Life Sciences 357544
382.15 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
Ideal for enzyme studies. Wheaton™ Dounce Tissue Grinders are designed to retain a high percentage of cell nuclei, mitochondria, and proteins in soft tissues or cell suspensions.
- Sample covers the pestle ball, preventing heat buildup by reducing friction against the mortar
- Mortar features large reservoir and pouring lip
- Supplied with “loose” and “tight” pestles
- Components are made of low extractable borosilicate glass
Order Info
- Tight pestle clearance: 0.0020 ±0.001 in. (0.05 ±0.025mm)
- Loose pestle clearance: 0.114 ±0.001 in. (0.114 ±0.025mm)
Specifications
0.5 oz. | |
Borosilicate Glass | |
Designed to retain a high percentage of cell nuclei, mitochondria and Proteins in soft tissues or cell suspensions | |
210mm | |
94mm | |
22mm |
15 mL | |
Dounce Tissue Grinder | |
8.26 in. | |
3.7 in. | |
0.86 in. | |
0.5 oz., 15mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction