Learn More
DWK Life Sciences Wheaton™ E-Z Decappers
With ergonomically designed handles for sealing and decapping standard laboratory vials
585.00 € - 1240.00 €
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
10785183
|
DWK Life Sciences
W225351 |
11 mm Seals |
585.00 €
Each |
Estimated Shipment: 25-03-2025
Log in to see stock. |
|||||
11521225
|
DWK Life Sciences
W225352 |
13 mm Seals |
1240.00 €
Each |
Estimated Shipment: 11-06-2025
Log in to see stock. |
|||||
10115822
|
DWK Life Sciences
W225353 |
20 mm Seals |
662.00 €
Each |
Estimated Shipment: 26-09-2025
Log in to see stock. |
|||||
11511225
|
DWK Life Sciences
W225350 |
8 mm Seals |
749.00 €
Each |
Estimated Shipment: 20-05-2025
Log in to see stock. |
|||||
Description
Wheaton E-Z Decappers are recommended for decapping standard laboratory vials.
- Several sizes of decapper are available, including those that remove seals 8mm, 11mm, 13mm and 20mm wide.
- The decappers come with visible size labels for easy identification.
- User comfort is preserved by an ergonomically designed, cushioned handle. Hand fatigue is reduced by the short handle and redesigned spring assembly.
- Decappers can be autoclaved, although autoclaving may change the appearance or reduce product life. They should not be autoclaved at greater than 121°C
Warranty
Two years