missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Glass 20-Slide Staining Dish with Removable Rack

Complete with dish, cover, and glass slide rack

Brand:  DWK Life Sciences 900200

Product Code. 10798324

  • 902.00 € / Pack of 6
Estimated Shipment: 25-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • With removable rack
  • Removable glass rack has a spring wire handle for manual staining of up to twenty horizontal slides
  • Bottom of rack is open for rapid immersion in and draining of stain
  • Holds slides measuring 3 × 1 in. (75 × 25mm), 3 × 1 1/2 in. (75 × 38mm), and 3 × 2 in. (75 × 51mm)
  • Made from soda-lime glass

Order Info

L x W x H:: 4.125 x 2.75 x 3.375 in. (105 x 70 x 85mm)

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Rectangular
Staining Dish
20
3.7 in.
2.9 in.
2.5 in.
6.4 cm
Soda Lime Glass
Removable Glass Slide Slotted Rack with Open Bottom
3.7 x 2.9 in. (9.5 x 7.6 cm)
95 mm
76 mm
64 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.