Learn More
DWK Life Sciences Wheaton™ Glass Sample Vials in Lab File With Caps Attached
Attached caps help maintain cleanliness
Brand:  DWK Life Sciences W224604
198.55 EUR valid until 2025-12-31
Use promo code "24350" to get your promotional price.
 View more versions of this product
Product Code. 10599502
  • 209.00 € / Pack of 72
Estimated Shipment: 10-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Attached caps help maintain cleanliness
Lab File with partitioned trays provides an easy way to store empty vials or to inventory samples.
  • Offer a variety of uses ranging from sample storage to freeze-drying
  • Fabricated from glass tubing for uniform sidewall and bottom thickness
  • Vials and caps are autoclavable
  • Amber vials made from borosilicate glass to protect light-sensitive products
Cap and Cap Liners
  • Solid top, black phenolic caps
  • 14B rubber-lined black phenolic screw cap for aqueous samples
  • PTFE/14B rubber-lined black phenolic screw cap for organic samples
Order Info
Replacements caps may be purchased separately.
Compliance
Amber vials conform to USP Type 1 requirements of light transmission.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PTFE/14B Rubber
28 mm
20 mL
Screw
Wheaton
Phenolic
Screw Cap
USP Type I
M-T File™ and Vial Rack
Amber
Borosilicate Glass
Low profile for greater stability and less dead volume above the sample, making it ideal for compound storage
5 dr.
20 mL
Autoclavable
60 mm
Vial