Learn More
DWK Life Sciences Wheaton™ Glass Sample Vials in Lab File without Caps
Packaged in partitioned trays, without caps
Brand:  DWK Life Sciences 224806
 View more versions of this product
Product Code. 10584805
  • 199.00 € / Pack of 200
Estimated Shipment: 17-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Partitioned trays provide an easy way to inventory samples or to store empty vials.
  • Autoclavable
  • Clear or Amber Vials
    • Clear vials made from low-extractable borosilicate glass that provides superior chemical resistance
    • Amber vials made from borosilicate glass that protects light-sensitive products
    Order Info
    Caps available separately
    Compliance
    Clear vials conform to USP Type 1 and ASTM™ E 438 Type 1, Class A requirements. Amber vials conform to USP Type 1 requirements for light transmission.
    TRUSTED_SUSTAINABILITY
    Specifications
    Specifications
    Clear
    Borosilicate Glass
    Screw
    Autoclavable
    70 mm
    Sample Vial in Lab File
    21 mm
    16 mL
    4.3 dr.
    Screw Cap
    ASTM E438, USP Type I, Type I, Class A
    M-T File™ and Vial Rack
    Product Suggestions
    Product Suggestions
    Videos
    SDS
    Documents
    Documents
    Certificates
    View accessory products