Learn More
DWK Life Sciences Wheaton™ Grinding Chamber Replacement Parts For Tapered Tissue Grinders With PTFE Pestles
Tapered tissue grinders replacement grinding chamber for fluoropolymer resin pestle
141.00 € - 166.00 €
Specifications
Material Borosilicate Glass
Product Type Grinding Chamber
For Use With (Application) Tapered tissue grinder with PTFE pestle
For Use With (Equipment) Tapered tissue grinder
View More Specs
Products 3
Product Code Brand Capacity Length (English) Tube Tube Diameter (English) Outside Price Quantity & Availability  
Product Code Brand Capacity Length (English) Tube Tube Diameter (English) Outside Price Quantity & Availability  
11758272
View Documents Promotion Details
DWK Life Sciences
358132
0.03 oz., 1mL 1.9 in. 0.43 in.
141.00 €
Pack of 2
Estimated Shipment: 16-05-2025
to see stock.
Add to basket
11738282
View Documents Promotion Details
DWK Life Sciences
358140
0.16 oz., 5mL 3.6 in. 0.51 in.
166.00 €
Pack of 2
Estimated Shipment: 16-05-2025
to see stock.
Add to basket
11758282
View Documents Promotion Details
DWK Life Sciences
358144
0.5 oz., 15mL 4.48 in. 0.7 in.
166.00 €
Pack of 2
Estimated Shipment: 16-05-2025
to see stock.
Add to basket
Description
Description
  • Grinders are available with glass or fluoropolymer resin pestles
  • Glass pestles are ideal for the homogenization of heart, muscle, lung, skin, and plant tissue
  • Fluoropolymer resin pestles are ideal for use with soft tissues such as brain and feature a stainless steel shaft with a 6.3mm (1/4 in.) diameter
  • Grinding chamber is made of low extractable borosilicate glass
  • The conical tip of the grinding chamber allows initial reduction, followed by passage through the cylindrical section for the final homogenization step
  • When pestle is “bottomed”- within the grinding chamber, the clearance is a uniform 0.102 to 0.152mm (0.004 to 0.006 in.)
Specifications
Specifications
Borosilicate Glass
Grinding Chamber
Tapered tissue grinder with PTFE pestle
Tapered tissue grinder