missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Hybridization Bottles
Shop All Wheaton Science Products Products
Click to view available options
Length (Metric):
75 mm
100 mm
150 mm
300 mm
Diameter (Metric) Inner:
35 mm
70 mm
Product Type:
Hybridization Bottle
Safety-coated Hybridization Bottle
Unit Size:
Each
Description
- Manufactured from heavy-wall borosilicate glass
- Safety-coated bottles with strong, slip-resistant plastisol covers reduce potential breakage and exposure to hazardous materials
- Polypropylene caps incorporate sonically bonded PTFE-fluorocarbon resin/silicone lining to prevent leakage
- Acrylic rack holds up to six hybridization bottles of different sizes for safe transport or storage
Informations sur la commande
Items come as one per case
Spécification
Spécification
| Certifications/Compliance | USP Type I |
| Closure Material | Polycarbonate |
| Closure Size | 45 mm |
| Coating | Plastisol |
| Diameter (English) Inner | 1.37 in. |
| Diameter (Metric) Inner | 35 mm |
| For Use With (Application) | Nucleic acid and protein blotting procedures that require a minimum of probe solution |
| Length (English) | 11.81 in. |
| Liner Material | PTFE or Silicone |
| Closure Type | Screw Cap |
| Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu