missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Hybridization Bottles

For use in nucleic acid and protein blotting procedures that require a minimum of probe solution

Brand:  DWK Life Sciences 805027

109.92 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 12437181

  • 121.00 € / Each
Estimated Shipment: 12-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Manufactured from heavy-wall borosilicate glass
  • Safety-coated bottles with strong, slip-resistant plastisol covers reduce potential breakage and exposure to hazardous materials
  • Polypropylene caps incorporate sonically bonded PTFE-fluorocarbon resin/silicone lining to prevent leakage
  • Acrylic rack holds up to six hybridization bottles of different sizes for safe transport or storage

Order Info

Items come as one per case

TRUSTED_SUSTAINABILITY
Specifications

Specifications

USP Type I
45 mm
1.37 in.
Nucleic acid and protein blotting procedures that require a minimum of probe solution
PTFE or Silicone
300 mm
Safety Coated Hybridization Bottle
Polycarbonate
Plastisol
35 mm
11.81 in.
Screw
Borosilicate Glass
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers