missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Hybridization Bottles

For use in nucleic acid and protein blotting procedures that require a minimum of probe solution

Brand:  DWK Life Sciences 805021

102.12 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 12871474

  • 113.00 € / Each
Estimated Shipment: 05-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Manufactured from heavy-wall borosilicate glass
  • Safety-coated bottles with strong, slip-resistant plastisol covers reduce potential breakage and exposure to hazardous materials
  • Polypropylene caps incorporate sonically bonded PTFE-fluorocarbon resin/silicone lining to prevent leakage
  • Acrylic rack holds up to six hybridization bottles of different sizes for safe transport or storage

Order Info

Items come as one per case

TRUSTED_SUSTAINABILITY
Specifications

Specifications

USP Type I
45 mm
1.37 in.
Nucleic acid and protein blotting procedures that require a minimum of probe solution
PTFE or Silicone
100 mm
Safety Coated Hybridization Bottle
Polycarbonate
Plastisol
35 mm
3.93 in.
Screw
Borosilicate Glass
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.