Learn More
DWK Life Sciences Wheaton™ Impellers for Celstir™ Spinner Flasks
Replacement for Double Sidearm Celstir and Jacketed Double Sidearm Celstir.
Brand:  DWK Life Sciences 356896
 View more versions of this product
Product Code. 12431421
  • 222.00 € / Each
Estimated Shipment: 10-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • This non-breakable stainless steel shaft, for replacing the glass shaft included with Celstir™ flasks, may be retrofitted to the existing Celstir flasks.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
125mL Celstir™ Spinner Flask
Top cap with liners, glass rod, magnet holder, magnet and stirring paddle
Impeller