Learn More
DWK Life Sciences™ Wheaton™ KeepIT™ Freezer Boxes
Provides an ideal method for batching and storing samples.
Brand:  DWK Life Sciences™ W651704-W
 View more versions of this product
Product Code. 13486626
  • 186.00 € / Pack of 10
Estimated Shipment: 07-11-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Six different colors match the colors of CryoELITE Cryogenic Vials, creating alternatives for batching and identifying groups of samples.
  • KeepIT-100 accommodates up to 100 internal threaded vials, KeepIT-81 accommodates 81 external threaded vials, and KeepIT-25 accommodates up to 25 internal or external threaded cryogenic vials; Vial sizes: 1.2 – 2mL
  • Openings in bottom facilitate scanning CryoELITE 2D Data Matrix Bar Code Inserts and allow for drainage of liquid nitrogen
  • Made from Eastman Tritan™ BPA-free, shatter-resistant resin
  • Standard footprint is compatible with liquid nitrogen storage shelves and freezer drawers
TRUSTED_SUSTAINABILITY
Specifications
Specifications
White
Square
Batching and Storing Samples
5.1 x 5.1 x 2 in. (13 x 13 x 5.2 cm)
13 cm
13 cm
5.2 cm
Copolyester Resin
Freezer Box
100 (1.2 to 2 mL Internal Threaded Cryogenic Vial)
5.1 in.
5.1 in.
2 in.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
DWK Life Sciences™ Wheaton™ KeepIT™ Freezer Boxes > KeepIT-100 Freezer Box; White