Learn More
DWK Life Sciences Wheaton™ Three-ball Snyder Column
For concentration of trace amounts of analyte in organic solvent
Brand:  DWK Life Sciences 384353
181.45 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
Product Code. 12431561
  • 208.00 € / Each
Currently Unavailable: We have no information when this product will be available
View alternative products
Explore more special offers
Description
Description
  • 24/40 clear seal joint
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Three-ball Snyder Column
24/40 clear seal joint
Concentration of trace amounts of analyte in organic solvent
ASTM E 438