Learn More
DWK Life Sciences Wheaton™ Evaporative Flask
For concentration of trace amounts of analyte in organic solvent
Brand:  DWK Life Sciences 297825
Product Code. 12488932
  • 143.00 € / Each
Estimated Shipment: 18-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • 24/40 top and 19/22 bottom clear seal joints
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Evaporative Flask
16.9 oz.
Concentration of trace amounts of analyte in organic solvent
500 mL
24/40 top and 19/22 bottom clearseal joints
ASTM E 438