Learn More
Wheaton™ Low-Density Extended Controlled Dropper Tips (15mm)
13mm extended controlled dropper tip made of LDPE (Low Density Polyethylene)
Brand:  Wheaton™ W242411-01-A
Product Code. 15162419
  • 337.00 € / Pack of 1000
Estimated Shipment: 10-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Extended controlled dropper tip dispenses one drop at a time
  • Average drop size is 44μL ±6μL
  • Case of 1000