Learn More
DWK Life Sciences Wheaton™ Low-Density Extended Controlled Dropper Tips
8mm extended controlled dropper tip made of LDPE (Low Density Polyethylene)
29.60 € - 260.00 €
Product Code | Brand | Color | Material | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Material | Price | Quantity & Availability | ||||
12436021
|
DWK Life Sciences
W242404-A |
Blue | Polyethylene |
258.00 €
Pack of 1000 |
Estimated Shipment: 31-03-2025
Log in to see stock. |
||||
10697333
|
DWK Life Sciences
242404 |
Blue | LDPE |
29.60 €
Pack of 100 |
Estimated Shipment: 31-03-2025
Log in to see stock. |
||||
12456021
|
DWK Life Sciences
W242405-A |
Gray | Polyethylene |
258.00 €
Pack of 1000 |
Estimated Shipment: 31-03-2025
Log in to see stock. |
||||
12438782
|
DWK Life Sciences
242405 |
Gray | LDPE |
58.65 €
Pack of 100 |
Estimated Shipment: 31-03-2025
Log in to see stock. |
||||
12476021
|
DWK Life Sciences
W242406-A |
Green | Polyethylene |
258.00 €
Pack of 1000 |
Estimated Shipment: 31-03-2025
Log in to see stock. |
||||
12448782
|
DWK Life Sciences
242406 |
Green | LDPE |
29.60 €
Pack of 100 |
Estimated Shipment: 31-03-2025
Log in to see stock. |
||||
10285702
|
DWK Life Sciences
W242401-01-A |
Natural | LDPE |
260.00 €
Pack of 1000 |
Estimated Shipment: 31-03-2025
Log in to see stock. |
||||
12416021
|
DWK Life Sciences
W242403-A |
Orange | Polyethylene |
258.00 €
Pack of 1000 |
Estimated Shipment: 31-03-2025
Log in to see stock. |
||||
12418782
|
DWK Life Sciences
242403 |
Orange | LDPE |
29.60 €
Pack of 100 |
Estimated Shipment: 31-03-2025
Log in to see stock. |
||||
12476011
|
DWK Life Sciences
W242401-A |
Pink | LDPE |
258.00 €
Pack of 1000 |
Estimated Shipment: 31-03-2025
Log in to see stock. |
||||
12488772
|
DWK Life Sciences
242401 |
Pink | LDPE |
29.60 €
Pack of 100 |
Estimated Shipment: 31-03-2025
Log in to see stock. |
||||
12496021
|
DWK Life Sciences
W242407-A |
Purple | Polyethylene |
258.00 €
Pack of 1000 |
Estimated Shipment: 31-03-2025
Log in to see stock. |
||||
11734209
|
DWK Life Sciences
242407 |
Purple | LDPE |
29.60 €
Pack of 100 |
Estimated Shipment: 31-03-2025
Log in to see stock. |
||||
10348773
|
DWK Life Sciences
W242402-A |
Red | Polyethylene |
258.00 €
Pack of 1000 |
Estimated Shipment: 31-03-2025
Log in to see stock. |
||||
10082643
|
DWK Life Sciences
242402 |
Red | LDPE |
29.60 €
Pack of 100 |
Estimated Shipment: 31-03-2025
Log in to see stock. |
||||
10488623
|
DWK Life Sciences
242410 |
White | LDPE |
29.60 €
Pack of 100 |
Estimated Shipment: 31-03-2025
Log in to see stock. |
||||
12416031
|
DWK Life Sciences
W242408-A |
Yellow | Polyethylene |
258.00 €
Pack of 1000 |
Estimated Shipment: 31-03-2025
Log in to see stock. |
||||
10438063
|
DWK Life Sciences
242408 |
Yellow | LDPE |
29.60 €
Pack of 100 |
Estimated Shipment: 31-03-2025
Log in to see stock. |
||||
Description
- Extended controlled dropper tip dispenses one drop at a time
- Average drop size is 44μL ±6μL