missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Lyophilized Stopper: 3-Leg Lyophilization Stopper

Very low adsorption of the preservatives present in aqueous parenteral solutions - Available in 2-Leg, 3-Leg and Igloo styles.

Brand:  DWK Life Sciences W224100-407

426.47 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.


Product Code. 10106047

  • 470.00 € / Pack of 1000
Estimated Shipment: 06-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Demonstrates a very low level of moisture adsorption
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications

Specifications

1.3 cm
Lyophilization
Autoclavable
Bromobutyl Rubber
2 cm
46 Durometer
Gray
3-Leg Lyophilization Stopper
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.