Learn More
DWK Life Sciences Wheaton™ M-T Vial File™ and Accessories
For compact and organized sample storage
Brand:  DWK Life Sciences 228783
 View more versions of this product
Product Code. 12453761
  • 42.75 € / Pack of 35
Explore more special offers
Description
Description
For compact and organized sample storage
  • Stores vials in clear polystyrene case
  • Foam inserts and partitions provide protection
  • With alphanumerical indexing card for identification
  • Index cards and partitions also available separately
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Storage and Transportation of Sample Vials
228778 Hinged Case
Index Card
35 Case