missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Magna-Flex™ Spinner Flasks
Produce high-yield cultures by maintaining even circulation while keeping microcarriers in suspension
Brand: DWK Life Sciences 356831
361.92 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Borosilicate glass flasks
- Two large side arms with screw-cap closures
- Flex-type, bulb-shaped glass impeller rotates from a fixed position above the liquid level around an indentation in the base of the flask
- Graduated
- Removable stainless-steel pin immobilizes the impeller during handling or decanting to prevent damage to cells or microcarriers
- Autoclavable flasks and impellers for selected sizes can also be ordered separately
- U.S. Patent No. 3,854,704
Specifications
250 mL | |
Flask with impeller | |
Autoclavable | |
51-440 |
Borosilicate Glass | |
33-430 | |
Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction