Learn More
DWK Life Sciences Wheaton™ Micro Kit and Accessories
WHEATON standard taper Clear-Seal™ grease-free joints provide smooth, unground surfaces, which seal without grease and reduce seizing and contamination from grease
Brand: DWK Life Sciences 109933
103.41 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Apparatus for a wide range of organic chemistry experiments at a lower price than purchasing the components separately
- Clear-Seal™ joints can replace ground units in any application and provide other important advantages over ground joints
- Superior in vacuum and radioactive applications
- Smooth surfaces resist contamination
- Cleaning is simple and thorough
- Interchangeable with ground joints
- All chemical changes and thermometer scales remain visible through the joints
- Clear-Seal joints have never been weakened by grinding and are formed from special wall tubing that is heavier than standard wall for extra strength
- Manufactured from WHEATON 33 low extractable borosilicate glass that conforms to ASTM E 438 Type I, Class A requirements
- Clear-Seal joints are produced to American Standard Taper dimensions and can be used interchangeably with any standard taper ground joints
Specifications
Wheaton micro kit | |
19/22 Clear-Seal™ joints | |
Claisen Adapter |