missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Omnispense™ Plus Peristaltic Pump Accessory Pump Heads

For use with Wheaton Omnispense Plus dispensing pump

Brand:  DWK Life Sciences W375023

 View more versions of this product

Product Code. 12452132

  • 626.00 € / Each
Estimated Shipment: 18-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Stackable pump heads (13-687-444 and -442) allow user to add up to two additional pumpheads, for a total of three pump heads per dispensing pump
  • Cassette pump head ideal for multichannel applications
  • Removable cassettes can be preloaded with tubing elements and changed without stopping the drive

†For tubing with bore sizes from 0.13 to 2.79mm.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Optional Pump Head for 1.6mm Wall Tubing
4
Pump Head
Omnispense™ plus peristaltic pump
1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.