Learn More
DWK Life Sciences Wheaton™ One-Piece Scrubber Absorber Tube
Modified version of catalog number 180020 for arsine analysis
Brand:  DWK Life Sciences 180024
Product Code. 12425482
  • 135.00 € / Each
Estimated Shipment: 15-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Eliminates the cumbersome ball and socket joint connecting the scrubber and absorber tubes which conforms to ASTM / USP and EPA specifications
  • The freestanding system has a standard taper 24/40 Clear-Seal joint
  • Manufactured from WHEATON 33 low extractable borosilicate glass that conforms to USP Type I and ASTM E 438 Type I, Class A requirements
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Scrubber/Absorber Tube
For arsenic analysis
Borosilicate Glass
1/Cs.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
DWK Life Sciences Wheaton™ One-Piece Scrubber Absorber Tube > Scrubber/Absorber Tube