missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Plastic Nongraduated Media Bottles
Shop All Wheaton Science Products Products
Click to view available options
Capacity (Metric):
125 mL
Description
Ideal for mixing and storing culture media, or for use with distilled water, analytical standards and reagents when fitted with a polyethylene-lined screw cap.
- Large opening, special lip and sloping shoulders for easy pouring, cleaning
- With polyethylene-lined caps
- Supplied with caps pre-attached and shrink-wrapped in convenient trays
Specifications
Specifications
Capacity (Metric) | 125 mL |
Color | Clear |
Diameter (English) | 2.08 in. |
For Use With (Application) | For distilled Water, analytical standards and reagents when fitted with a Polyethylene lined screw cap |
Material | PET |
Quantity | 24 Cs. |
Sterility | Sterile |
Neck Finish | 33-430 |
Graduated | No |
Shape | Square |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction