missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Polypropylene Caps for Wheaton Dropping Bottles - Yellow

Product Code. 12448842
Click to view available options
Neck Finish:
13-425
15-415
20-410
8-425
For Use With (Equipment):
Dropping bottle
Plastic dropping bottle
No. per Case:
100 Cs.
1000 Cs.
This item is not returnable. View return policy
This item is not returnable. View return policy

Screw cap made of radiation grade polypropylene

  • Packaged in case of 100 or 1000
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Material Polypropylene
No. per Case 100 Cs.
Neck Finish 20-410
Certifications/Compliance ISO 9001
Product Type Polypropylene Cap
For Use With (Equipment) Dropping bottle
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'