missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Polypropylene Caps for Wheaton Dropping Bottles - Yellow
Click to view available options
Neck Finish:
13-425
15-415
20-410
8-425
For Use With (Equipment):
Dropping bottle
Plastic dropping bottle
No. per Case:
100 Cs.
1000 Cs.
Description
- Packaged in case of 100 or 1000
Specifications
Specifications
Color | Yellow |
Material | Polypropylene |
No. per Case | 1000 Cs. |
Neck Finish | 15-415 |
Certifications/Compliance | ISO 9001 |
Product Type | Polypropylene Cap |
For Use With (Equipment) | Plastic dropping bottle |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'