Learn More
DWK Life Sciences Wheaton™ Replacement O-Rings for Dry-Seal Vacuum Desiccator
For use with the Wheaton™ Dry-Seal Vacuum Desiccator.
Brand:  DWK Life Sciences 366022
Product Code. 12431521
  • 47.65 € / Each
Estimated Shipment: 22-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Constructed of slicone rubber
TRUSTED_SUSTAINABILITY
Specifications
Specifications
100 mm
Silicone Rubber
For dry-seal vacuum desiccator
Replacement O-ring
Createing an airtight seal when vacuum is applied to dessicator
4 in.