missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Screw Neck Glass Diagnostic Bottles
Shop All Wheaton Science Products Products
Click to view available options
Capacity: 10cc
Description
- Autoclavable
- Amber-colored glass
- Can by used for lyophilization
- Alternative to serum bottles and vials
- Fits with I-Loc™ closure or screw cap with thin flange stopper
Specifications
Specifications
Capacity | 0.34 oz., 10 mL |
Capacity (Metric) | 10 mL |
Capacity (English) | 0.34 oz. |
Color | Amber |
Diameter (English) | 0.9 in. |
Material | Glass |
Certifications/Compliance | USP Type I |
Height (Metric) | 61 mm |
Height (English) | 2.4 in. |
Closure Size | 20-400 mm |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction