missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ 47mm 100mL Glass Funnel

Brand:  DWK Life Sciences 419410

Product Code. 12604833

  • 95.55 € / Each
Estimated Shipment: 14-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

TRUSTED_SUSTAINABILITY
Specifications

Specifications

100 mL
USP Type I, ASTM E438 Type I, Class A
47mm
40 to 60μm
Borosilicate Glass
Designed to handle up to 1000mL of liquid for analysis of particulate or microbiological contamination
Coarse
Funnel
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers