Learn More
DWK Life Sciences Wheaton™ Arsine Generator With Clear–Seal Joint And 24/40 Standard Taper
Engineered for arsine analysis using the silver diethyldithiocarbamate (SDDC) colorimetric method
Brand:  DWK Life Sciences 180020
Product Code. 12495472
  • 194.00 € / Each
Estimated Shipment: 17-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Manufactured from WHEATON 33 low extractable borosilicate glass
Manufactured from WHEATON 33 low extractable borosilicate glass that conforms to USP Type I and ASTM E 438 Type I, Class A requirements; Reference: £Standard Test Method for Arsenic in Water,£ASTM D2972
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Arsine Generator
Borosilicate Glass
For arsine analysis using SDDC method
24/40
USP Type I and ASTM E438 Type I, Class A
125mL erlenmeyer flask scrubber tube, socket joint, size 12 stainless steel ball joint clamp