missing translation for 'onlineSavingsMsg'
Learn More
DWK Life Sciences Wheaton™ Serum Bottles and Vials Product Code.: 10632973
In Stock
to see stock.
Add to basket

DWK Life Sciences Wheaton™ Serum Bottles and Vials

Product Code. 10632973 Shop All Wheaton Science Products Products
30 mL, Clear
Click to view available options
Capacity (Metric):
2 mL
5 mL
10 mL
20 mL
30 mL
50 mL
60 mL
100 mL
125 mL
Color:
Amber
Clear
This item is not returnable. View return policy

Product Code. 10632973

Brand: DWK Life Sciences 223743

In Stock
to see stock.
Edge
Add to basket
Add to basket
This item is not returnable. View return policy

These bottles have exceptional wall strength and superior heat and chemical tolerances

Ideal for long- and short-term sample storage and vaccine/injectable drug containers.

  • Clear borosilicate glass conforms to USP Type 1 requirements
  • Amber borosilicate glass conforms to USP Type 1 requirements
  • Shrink wrapped partitioned modules reduce breakage

Compliance

Meet requirements for Class A of the United States Pharmacopeia for Type I glass.

TRUSTED_SUSTAINABILITY

Specifications

Capacity 1.01 oz., 30 mL
Capacity (Metric) 30 mL
Capacity (English) 1.01 oz.
Color Clear
Diameter (English) 1.45 in.
Material Borosilicate Glass
Height (Metric) 63 mm
Height (English) 2.48 in.
Diameter (Metric) 37 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DWK Life Sciences Wheaton™ Serum Bottles and Vials >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.