missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Serum Vials
Available as clear and amber vials
Brand: DWK Life Sciences 223696
This item is not returnable.
View return policy
Description
Available as clear and amber vials
Glass can withstand the challenges of reduced pressure and extreme temperature variations. Protects against changes in pH and maintain the purity of the contents.
- Clear vials made from low extractable borosilicate glass tubing to ensure uniform wall thickness
- Amber vials made from borosilicate glass conforms to USP Type 1 requirements to protect light-sensitive samples
- Withstand extreme temperature variations
- Necks finished for aluminum crimp seals
- Blowback feature of neck and specially designed bottom radius for lyophilization applications
- Packaged in shrink-wrapped modules to reduce particulate contamination
Lyophilization, packaging, and sampling
Compliance
Low extractable borosilicate glass meets ASTM™ Type 1, Class A and USP Type 1 standards

Specifications
0.51 in. | |
Amber | |
Borosilicate Glass | |
For lyophilization, packaging, sampling and freeze drying applications | |
Autoclavable | |
50 mm |
13 mm | |
24 mm | |
10 mL | |
2.7 dr. | |
Crimp Cap | |
ASTM E438, USP Type I, Type I, Class A |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
WHE026 Catalogue 2012 FINAL LOWRES
File size: 14.8 MB
WHE026 Catalogue 2012 FINAL LOWRES
File size: 14.8 MB
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DWK Life Sciences Wheaton™ Serum Vials > Amber; 10mL
Spot an opportunity for improvement?Share a Content Correction