Learn More
DWK Life Sciences™ Wheaton™ Serum Vial

Serum vials are converted from Type I borosilicate glass tubing.
Brand: DWK Life Sciences™ WUBB2MLA-T-EP
190.88 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Vials conform to the hydrolytic resistance requirements outlined in USP 660 and EP 3.2.1
- Shrink-wrapped modules reduce particulate contamination
- Special design adds strength for freeze-drying applications

Specifications
0.28 in. | |
7 mm | |
13 mm | |
No | |
2 mL | |
0.07 oz. | |
Crimp Cap | |
225 Vials/Pk. | |
Serum Vial |
0.51 in. | |
Amber | |
Borosilicate Glass | |
Non-sterile | |
1.26 in. | |
Autoclavable | |
32 mm | |
USP Type I | |
No |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.