missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences™ Wheaton™ Serum Vial
GREENER_CHOICE

Serum vials are converted from Type I borosilicate glass tubing.

Brand:  DWK Life Sciences™ WUBB3MLA-T-EP

 View more versions of this product

Product Code. 18265435

  • 192.00 € / Pack of 169
Estimated Shipment: 23-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Vials conform to the hydrolytic resistance requirements outlined in USP 660 and EP 3.2.1
  • Shrink-wrapped modules reduce particulate contamination
  • Special design adds strength for freeze-drying applications
TRUSTED_SUSTAINABILITY
Specifications

Specifications

0.28 in.
7 mm
13 mm
No
3 mL
0.1 oz.
Crimp Cap
169 Vials/Pk.
Serum Vial
0.51 in.
Amber
Borosilicate Glass
Non-sterile
1.48 in.
Autoclavable
37.7 mm
USP Type I
No
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DWK Life Sciences™ Wheaton™ Serum Vial > Serum Vials; Color: Amber; Volume (Metric): 3 mL; Quantity: 169 Vials/Pk.

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.