Learn More
DWK Life Sciences Wheaton™ Soxhlet Extraction Apparatus With Standard Taper Ground Joints
For continuous extraction of analytes from a solid into an organic solvent
Brand:  DWK Life Sciences 416000
 View more versions of this product
Product Code. 12674823
  • 677.00 € / Each
Estimated Shipment: 07-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Manufactured from low extractable borosilicate glass
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Soxhlet Extraction Apparatus
4.22 oz.
Standard taper ground joints, 15mL graduated funnel, anodized aluminum clamp, No. 5 silicone stopper
ASTM E 438
24/40
125 mL
Borosilicate Glass
Used for continuous extraction of analytes from a solid into an organic solvent
34/45