missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Spin Vanes for V-Vials

Magnetic stir bar for use with NextGen™ V Vials™

117.00 € - 122.00 €

Specifications

Material PTFE
Quantity 6
Shape Triangle
View More Specs

Products 2
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
10226863
View Documents
DWK Life Sciences
903061
Fit 0.3 and 1mL V-Vials
117.00 €
Pack of 6
Estimated Shipment: 27-01-2025
to see stock.
Add to basket
 
10168421
View Documents
DWK Life Sciences
903063
Fit 2, 3, 5, and 10mL V-Vials
122.00 €
Pack of 6
Estimated Shipment: 04-03-2025
to see stock.
Add to basket
 
Description

Description

  • Made from PTFE
  • Place stir bar in vial and use with magnetic stirrer
Specifications

Specifications

PTFE
6
Triangle
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.