missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Stainless Steel Scoop

Ideal for sampling powders, slurries, soils and more

Brand:  DWK Life Sciences 885540

Product Code. 12427321

  • 203.00 € / Pack of 4
Estimated Shipment: 29-10-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Sturdy 302 stainless-steel construction
  • Length: 24cm (9.5 in.)
  • Scoop Depth: 2.54cm (1 in.)

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Scoop
Ideal for sampling powders, slurries, soils and more
24 cm
Stainless Steel
9.5 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers