missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Standard Opening Glass Vials

Made with Type I Borosilicate glass

Brand:  DWK Life Sciences 223682-01

 View more versions of this product

Product Code. 12442591

  • 280.00 € / Pack of 1000
Estimated Shipment: 06-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • 1.5mL (0.05 oz.)
  • 12 dia. x 32mm H (0.47 dia. x 1.26 in. H)
  • 11mm (0.43 in.) Crimp Top

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Clear
Borosilicate Glass
1.25 in.
Crimp Cap
1000 Cs.
Yes
12 mm
1.5 mL
0.05 oz.
32 mm
11 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers