missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences™ Wheaton™ Thin Flange Stopper for Serum Bottles and Vials

Provide airtight covers for your serum bottles and vials with this thin flange stopper.

Brand:  DWK Life Sciences™ 224100-203

197.85 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 11561105

  • 215.00 € / Pack of 300
Estimated Shipment: 19-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Flange-type stoppers are secured with aluminum crimp seals
  • Slotted and three-prong plug stoppers are ideal for freeze-dry applications
TRUSTED_SUSTAINABILITY
Specifications

Specifications

3-leg
Lyophilization
300 Stoppers/Pk.
Gray
Thin Flange Stopper
Non-sterile
2 cm
Serum Bottles and Vials 1.3 cm I.D. and 2 cm O.D.
Autoclavable
Chlorobutyl Rubber
Ribbed Cone
20
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.