missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Stoppers for Serum Bottles and Vials: 3-Leg Lyophilization Stopper
Provide airtight covers
Brand: DWK Life Sciences W224100-202
259.35 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
Provide airtight covers
- Flange-type stoppers are secured with aluminum crimp seals
- Slotted and three-prong plug stoppers are for freeze-dry applications
Specifications
2 cm | |
Fits 1.3cm I.D., 2cm O.D. mouth of serum bottles and vials | |
Autoclavable | |
Butyl Rubber |
Lyophilization | |
50 Durometer | |
Gray | |
3-Leg Lyophilization Stopper |