missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Stoppers for Serum Bottles and Vials: 2-Leg Stoppers

Provide airtight covers

Brand:  DWK Life Sciences W224100-093

171.38 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 11764189

  • 222.00 € / Pack of 1000
Estimated Shipment: 09-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Provide airtight covers

  • Flange-type, straight plug stoppers are secured with aluminum crimp seals (06-406-11B)
  • Three-prong plug stoppers are for freeze-dry applications (06-447E, 06-451-56)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

7 mm
Lyophilization
50 Durometer
Gray
2-Leg Lyophilization Stopper; Butyl Rubber
13 mm
Fits 7mm I.D., 13mm O.D. mouth of serum bottles and vials
Autoclavable
Butyl Rubber
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.