Learn More
DWK Life Sciences Wheaton™ Straight Plug Rubber Stoppers
Provides maximum sealing for vials with straight-wall glass finishes. Wheaton™ Straight Plug Rubber Stoppers are manufactured from the highest quality raw materials.
Brand:  DWK Life Sciences 224100-175
1479.99 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 10403085
  • 2270.00 € / Pack of 1000
Estimated Shipment: 05-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Autoclavable
  • Available in a variety of sizes/colors
  • Manufactured to yield exceptional dimensional stability
TRUSTED_SUSTAINABILITY
Specifications
Specifications
20 mm
FEP
Vacule Ampules
Straight Plug Stopper
Gray