missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Tenbroeck Tissue Grinder

With expanded reservoir and pouring lip

244.00 € - 450.00 €

Specifications

Material Borosilicate Glass
Product Type Tenbroeck Tissue Grinder
View More Specs

Products 5
Product Code Brand Capacity Length (English) Length (English) Tube Tube Diameter (English) Outside Price Quantity & Availability  
Product Code Brand Capacity Length (English) Length (English) Tube Tube Diameter (English) Outside Price Quantity & Availability  
10698643
View Documents
DWK Life Sciences
357428
1.35 oz., 40mL 12.5 in. 5.5 in. 1.25 in.
450.00 €
Pack of 2
Estimated Shipment: 07-04-2025
to see stock.
Add to basket
 
10428341
View Documents
DWK Life Sciences
357421
0.03 oz., 1mL 5.51 in. 1.8 in. 0.43 in.
244.00 €
Pack of 2
Estimated Shipment: 24-01-2025
to see stock.
Add to basket
 
10688643
View Documents
DWK Life Sciences
357422
0.06 oz., 2mL 5.51 in. 1.9 in. 0.43 in.
313.00 €
Pack of 2
Estimated Shipment: 09-05-2025
to see stock.
Add to basket
 
10198611
View Documents
DWK Life Sciences
357424
0.23 oz., 7mL 7.4 in. 3.2 in. 0.62 in.
321.00 €
Pack of 2
Estimated Shipment: 27-01-2025
to see stock.
Add to basket
 
10549930
View Documents
DWK Life Sciences
357426
0.5 oz., 15mL 9.8 in. 3.7 in. 0.86 in.
345.00 €
Pack of 2
Estimated Shipment: 07-04-2025
to see stock.
Add to basket
 
Description

Description

With expanded reservoir and pouring lip

  • Precision manufactured from Wheaton-33™ low extractable borosilicate glass
  • Can be operated by hand or slow-speed motor
  • Hollow handle permits packing with ice to reduce heat buildup
  • Clearance between pestle and tube is approximately 0.12mm

Specifications

Specifications

Borosilicate Glass
Tenbroeck Tissue Grinder
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.