missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Trypsinizing Flasks

Deep fluted sides enhance agitation

Brand:  DWK Life Sciences 355392

65.73 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 10306952

  • 71.90 € / Each
Estimated Shipment: 10-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Deep fluted sides enhance agitation

  • Manufactured from low-extractable borosilicate glass
  • Excellent quality and low extractables
  • Deep-skirted rubber-lined screw caps
  • Available graduated or ungraduated

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Style: Erlenmeyer Trypsinizing
Borosilicate Glass
430
No
24 mm
35 mL
Used for forming cell suspensions through Trypsin digestion of connective Proteins
Autoclavable
Screw
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers