Learn More
DWK Life Sciences™ Wheaton™ Vacule™ Lyophilization Ampules
For small volume samples
Brand: DWK Life Sciences™ W651446
This item is not returnable.
View return policy
Description
- Made from Type I borosilicate glass
- Available with heat or stopper seal
Specifications
Borosilicate Glass (Type I) | |
0.472 in. | |
3.3 in. |
1.2 cm | |
86 mm | |
0.3 in. |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
DWK Life Sciences™ Wheaton™ Vacule™ Lyophilization Ampules > 2 Vacule; Dia. x H: 0.4 x 3.3 in. (12 x 86mm); O.D. at top: 0.3 in. (10mm) approx.; 567/Cs.
Spot an opportunity for improvement?Share a Content Correction